Product Details

SNP ID
rs3751911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82657767 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACACATTCGCAAGCAGCATTCGCC[A/G]AGAGGAACCGGGATCTGAGATGCAT
Phenotype
MIM: 609226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB40B PubMed Links

Gene Details

Gene
RAB40B
Gene Name
RAB40B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006822.2 1171 UTR 3 NP_006813.1
XM_005256334.4 1171 UTR 3 XP_005256391.1
XM_005256335.3 1171 Intron XP_005256392.1
XM_006722271.3 1171 UTR 3 XP_006722334.1
XM_011523528.2 1171 UTR 3 XP_011521830.1
XM_017024042.1 1171 Intron XP_016879531.1
XM_017024043.1 1171 UTR 3 XP_016879532.1
Gene
WDR45B
Gene Name
WD repeat domain 45B
There are no transcripts associated with this gene.

View Full Product Details