Product Details

SNP ID
rs3760901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4010344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTGGGTGGGGCAGGGGCTCCC[A/G]GGGTGCGAGGATGTCAGCAGGTTCC
Phenotype
MIM: 605989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIAS4 PubMed Links

Gene Details

Gene
PIAS4
Gene Name
protein inhibitor of activated STAT 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015897.3 379 Intron NP_056981.2
XM_011528060.2 379 UTR 5 XP_011526362.1
XM_017026868.1 379 Intron XP_016882357.1

View Full Product Details