Product Details
- SNP ID
-
rs28937580
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:56594326 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCAGCACTATCTCCACATCCGC[C/T]CGGCACCCAGCGACAACCTGCCCCT
- Phenotype
-
MIM: 602991
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NOG
PubMed Links
Gene Details
- Gene
- NOG
- Gene Name
- noggin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005450.4 |
628 |
Missense Mutation |
CCG,TCG |
P35S |
NP_005441.1 |
View Full Product Details