Product Details

SNP ID
rs144469564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:25427013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTATTACAGCGATGATCTCATAA[C/T]ATCACTGAAATGACAGATGAAAAAA
Phenotype
MIM: 152760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNRH1 PubMed Links

Gene Details

Gene
GNRH1
Gene Name
gonadotropin releasing hormone 1
There are no transcripts associated with this gene.

Gene
KCTD9
Gene Name
potassium channel tetramerization domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017634.3 Intron NP_060104.2

View Full Product Details