Product Details

SNP ID
rs4940736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:58678199 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCAACATTATCACTGAATTTTAAA[A/T]CTTGGGTAACAGTATAAATGCTATG
Phenotype
MIM: 604860
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927322 PubMed Links

Gene Details

Gene
LOC101927322
Gene Name
uncharacterized LOC101927322
There are no transcripts associated with this gene.

Gene
LOC102724745
Gene Name
uncharacterized LOC102724745
There are no transcripts associated with this gene.

Gene
MALT1
Gene Name
MALT1 paracaspase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006785.3 Intron NP_006776.1
NM_173844.2 Intron NP_776216.1
XM_011525794.1 Intron XP_011524096.1

View Full Product Details