Product Details

SNP ID
rs192136138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27919661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGACACCTCACCTCCATAGTCC[A/T]CATAAAAGACTCCTGACAGGTTCTG
Phenotype
MIM: 606876
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR3199-1 PubMed Links

Gene Details

Gene
MIR3199-1
Gene Name
microRNA 3199-1
There are no transcripts associated with this gene.

Gene
MIR3199-2
Gene Name
microRNA 3199-2
There are no transcripts associated with this gene.

Gene
PITPNB
Gene Name
phosphatidylinositol transfer protein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284277.1 Intron NP_001271206.1
NM_001284278.1 Intron NP_001271207.1
NM_012399.4 Intron NP_036531.1
XM_011530052.2 Intron XP_011528354.1
XM_017028707.1 Intron XP_016884196.1
Gene
TTC28-AS1
Gene Name
TTC28 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details