Product Details
- SNP ID
-
hCV27854787
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:109095707 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGGACTTTTTCCAGTGACTTGC[C/G]TGGGAAGGACTAGTCCAGCTGCCCA
- Phenotype
-
MIM: 606103
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CEP57L1
PubMed Links
Gene Details
- Gene
- CEP57L1
- Gene Name
- centrosomal protein 57 like 1
There are no transcripts associated with this gene.
- Gene
- SESN1
- Gene Name
- sestrin 1
There are no transcripts associated with this gene.
View Full Product Details