Product Details

SNP ID
rs4234696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50622310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTCTAACTGTGGCATTCAAAG[C/T]CTCTGCAGTCCAGCCCACCTCCCTT
Phenotype
MIM: 602130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPKAPK3 PubMed Links

Gene Details

Gene
MAPKAPK3
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243925.1 Intron NP_001230854.1
NM_001243926.1 Intron NP_001230855.1
NM_004635.4 Intron NP_004626.1

View Full Product Details