Product Details
- SNP ID
-
rs4880266
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:132540863 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCCGGTCACAGTTCCGAGGAGGAAG[G/A]TGTTTGAATCTGAAGCAGATATCCC
- Phenotype
-
MIM: 600106
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INPP5A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79919904] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INPP5A
- Gene Name
- inositol polyphosphate-5-phosphatase A
- Gene
- LOC107984282
- Gene Name
- uncharacterized LOC107984282
There are no transcripts associated with this gene.
View Full Product Details