Product Details

SNP ID
rs4455803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:55780594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTATTTTGTGGGGGCATATTCCA[C/G]TGTTATATCAGTATCCTGTTTTCCA
Phenotype
MIM: 606461
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TGS1 PubMed Links

Gene Details

Gene
TGS1
Gene Name
trimethylguanosine synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317902.1 Intron NP_001304831.1
NM_024831.7 Intron NP_079107.6
Gene
TMEM68
Gene Name
transmembrane protein 68
There are no transcripts associated with this gene.

View Full Product Details