Product Details

SNP ID
rs4713842
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35226855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCCTTCAAGTATAGCTGTGTGT[A/G]TAGCACCTATTGTTAGAGCTAGGGC
Phenotype
MIM: 614708
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SCUBE3 PubMed Links

Gene Details

Gene
SCUBE3
Gene Name
signal peptide, CUB domain and EGF like domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303136.1 Intron NP_001290065.1
NM_152753.3 Intron NP_689966.2
XM_005248943.1 Intron XP_005249000.1
XM_005248947.1 Intron XP_005249004.1
XM_011514404.2 Intron XP_011512706.1

View Full Product Details