Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001304361.1 | 4467 | UTR 3 | NP_001291290.1 | ||
NM_152515.4 | 4467 | UTR 3 | NP_689728.3 | ||
XM_011510666.2 | 4467 | Intron | XP_011508968.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_011512262.2 | 4467 | Intron | XP_011510564.1 | ||
XM_011512265.2 | 4467 | Intron | XP_011510567.1 | ||
XM_017005474.1 | 4467 | Intron | XP_016860963.1 | ||
XM_017005475.1 | 4467 | Intron | XP_016860964.1 | ||
XM_017005476.1 | 4467 | Intron | XP_016860965.1 | ||
XM_017005477.1 | 4467 | Intron | XP_016860966.1 | ||
XM_017005478.1 | 4467 | Intron | XP_016860967.1 |