Product Details
- SNP ID
-
rs6819838
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:5014833 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTAATAAAAACATACTATTGTGCA[A/T]GTTCTCAAAATAGTTGTTCATAAAA
- Phenotype
-
MIM: 607930
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CYTL1
PubMed Links
Gene Details
- Gene
- CYTL1
- Gene Name
- cytokine like 1
View Full Product Details