Product Details
- SNP ID
-
rs8111318
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:799066 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCAACTTACTGGAAAGCACACAG[C/G]AGCCCTTGAACCCTCTCAGCGGCAT
- Phenotype
-
MIM: 600693
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4745
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113844239,rs77100342] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR4745
- Gene Name
- microRNA 4745
There are no transcripts associated with this gene.
- Gene
- PTBP1
- Gene Name
- polypyrimidine tract binding protein 1
View Full Product Details