Product Details

SNP ID
rs7943454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:24500120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAATCCCATAATTTAATCACAG[C/T]GAACATCAATGGCCCCCAATCAGAG
Phenotype
MIM: 608178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LUZP2 PubMed Links

Gene Details

Gene
LUZP2
Gene Name
leucine zipper protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009909.3 Intron NP_001009909.2
NM_001252008.1 Intron NP_001238937.1
NM_001252010.1 Intron NP_001238939.1
XM_011520056.2 Intron XP_011518358.1
XM_017017648.1 Intron XP_016873137.1
XM_017017649.1 Intron XP_016873138.1
XM_017017650.1 Intron XP_016873139.1
XM_017017651.1 Intron XP_016873140.1

View Full Product Details