Product Details

SNP ID
rs7925604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71930210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCTTGCACATGCCAGCATATGG[A/G]CAAAGACATCACAGGAATAGGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100133315 PubMed Links

Gene Details

Gene
LOC100133315
Gene Name
transient receptor potential cation channel, subfamily C, member 2-like
There are no transcripts associated with this gene.

Gene
RNF121
Gene Name
ring finger protein 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300926.1 Intron NP_001287855.1
NM_018320.4 Intron NP_060790.2
XM_006718629.1 Intron XP_006718692.1
XM_006718630.1 Intron XP_006718693.1
XM_006718632.1 Intron XP_006718695.1
XM_011545150.1 Intron XP_011543452.1

View Full Product Details