Product Details

SNP ID
rs7957187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56042327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGAGAAACAGGAGATCCGAGCGG[C/G]GCCTTCCTGGAGGCTGCCGGTGCGG
Phenotype
MIM: 606239 MIM: 603701
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IKZF4 PubMed Links

Gene Details

Gene
IKZF4
Gene Name
IKAROS family zinc finger 4
There are no transcripts associated with this gene.

Gene
RPS26
Gene Name
ribosomal protein S26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029.3 Intron NP_001020.2

View Full Product Details