Product Details

SNP ID
rs7309325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111411711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGTGGACCCGGTTGTGTTCTGA[G/T]CTTGGGGTCCTGAGATGCAGACACC
Phenotype
MIM: 605093
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SH2B3 PubMed Links

Gene Details

Gene
SH2B3
Gene Name
SH2B adaptor protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291424.1 Intron NP_001278353.1
NM_005475.2 Intron NP_005466.1
XM_005253818.4 Intron XP_005253875.1
XM_005253819.4 Intron XP_005253876.1
XM_006719180.3 Intron XP_006719243.1
XM_011537719.2 Intron XP_011536021.1
XM_011537720.2 Intron XP_011536022.1
XM_011537721.2 Intron XP_011536023.1

View Full Product Details