Product Details

SNP ID
rs6531933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:86825341 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCACAGGGGCAGCACATGCAAGT[C/T]TGGATTGAAAAAAAAGTTGGTAAGT
Phenotype
MIM: 613366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC10A6 PubMed Links

Gene Details

Gene
SLC10A6
Gene Name
solute carrier family 10 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197965.2 Intron NP_932069.1

View Full Product Details