Product Details
- SNP ID
-
rs7028425
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:35606741 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGCACAGTCTTACCCCTATGACCT[C/G]TGTGATCCTCGGGAGTGTCCCCTAG
- Phenotype
-
MIM: 107272
MIM: 601782
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CD72
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs577133285] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD72
- Gene Name
- CD72 molecule
There are no transcripts associated with this gene.
- Gene
- MIR4667
- Gene Name
- microRNA 4667
There are no transcripts associated with this gene.
- Gene
- TESK1
- Gene Name
- testis-specific kinase 1
View Full Product Details