Product Details
- SNP ID
-
rs7161271
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:22603137 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTTTGCAGGATTCTTGCTAAGACT[A/G]AGCTAGACATTTGGAAGACAGGCCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ABHD4
PubMed Links
Gene Details
- Gene
- ABHD4
- Gene Name
- abhydrolase domain containing 4
View Full Product Details