Product Details

SNP ID
rs8118442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35522646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAAAAGGCTTCGGCTTGGTCTT[C/G]GTCTCCACCTCCTACCCTGCACCCA
Phenotype
MIM: 609689
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C20orf173 PubMed Links

Gene Details

Gene
C20orf173
Gene Name
chromosome 20 open reading frame 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145350.1 Intron NP_001138822.1
XM_006723710.2 Intron XP_006723773.1
XM_011528577.1 Intron XP_011526879.1
Gene
CEP250
Gene Name
centrosomal protein 250
There are no transcripts associated with this gene.

View Full Product Details