Product Details
- SNP ID
-
rs8059926
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1711318 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGGCCCCTCGTTCTCTGCTCTTA[G/T]TGACCTTGACCATTGCGACAACACA
- Phenotype
-
MIM: 605431
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HN1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8055429] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HN1L
- Gene Name
- hematological and neurological expressed 1-like
There are no transcripts associated with this gene.
- Gene
- MAPK8IP3
- Gene Name
- mitogen-activated protein kinase 8 interacting protein 3
View Full Product Details