Product Details

SNP ID
rs7470145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122919905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAATCTCCATAATTTTCAAACTT[A/G]AAGTGAAGGAGATCTGATCTTTCAG
Phenotype
MIM: 605976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB26 PubMed Links

Gene Details

Gene
ZBTB26
Gene Name
zinc finger and BTB domain containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304363.1 113 Silent Mutation TTC,TTT F10F NP_001291292.1
NM_001304364.1 113 Silent Mutation TTC,TTT F10F NP_001291293.1
NM_020924.3 113 Silent Mutation TTC,TTT F10F NP_065975.1
Gene
ZBTB6
Gene Name
zinc finger and BTB domain containing 6
There are no transcripts associated with this gene.

View Full Product Details