Product Details

SNP ID
rs7962715
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:101597779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTGGAGAACTATGTTATTTGTA[C/T]TGTCTATTCCAGATAAAACTTACCT
Phenotype
MIM: 160794
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MYBPC1 PubMed Links

Gene Details

Gene
MYBPC1
Gene Name
myosin binding protein C, slow type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254718.1 Intron NP_001241647.1
NM_001254719.1 Intron NP_001241648.1
NM_001254720.1 Intron NP_001241649.1
NM_001254721.1 Intron NP_001241650.1
NM_001254722.1 Intron NP_001241651.1
NM_001254723.1 Intron NP_001241652.1
NM_002465.3 Intron NP_002456.2
NM_206819.2 Intron NP_996555.1
NM_206820.2 Intron NP_996556.1
NM_206821.2 Intron NP_996557.1
XM_005268876.4 Intron XP_005268933.1
XM_006719405.3 Intron XP_006719468.1
XM_006719406.3 Intron XP_006719469.1
XM_006719407.3 Intron XP_006719470.1
XM_006719408.3 Intron XP_006719471.1
XM_006719409.3 Intron XP_006719472.1
XM_006719410.3 Intron XP_006719473.1
XM_006719411.3 Intron XP_006719474.1
XM_017019315.1 Intron XP_016874804.1
XM_017019316.1 Intron XP_016874805.1
XM_017019317.1 Intron XP_016874806.1
XM_017019318.1 Intron XP_016874807.1
XM_017019319.1 Intron XP_016874808.1
XM_017019320.1 Intron XP_016874809.1
XM_017019321.1 Intron XP_016874810.1
XM_017019322.1 Intron XP_016874811.1

View Full Product Details