Product Details

SNP ID
rs7916991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69135617 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTTTTTAAAAATTACATTTTGG[A/C]ATTGCTGGAAACCATACGTAGACCT
Phenotype
MIM: 605506
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VPS26A PubMed Links

Gene Details

Gene
VPS26A
Gene Name
VPS26, retromer complex component A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035260.2 Intron NP_001030337.1
NM_001318944.1 Intron NP_001305873.1
NM_001318945.1 Intron NP_001305874.1
NM_001318946.1 Intron NP_001305875.1
NM_004896.4 Intron NP_004887.2

View Full Product Details