Product Details

SNP ID
rs5910613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119549663 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGAAGCATAAGAGCATGGCTCC[A/G]GCATTGGGCAAGGGCCTTCGTGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf56 PubMed Links

Gene Details

Gene
CXorf56
Gene Name
chromosome X open reading frame 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170569.1 Intron NP_001164040.1
NM_001170570.1 Intron NP_001164041.1
NM_022101.3 Intron NP_071384.1

View Full Product Details