Product Details

SNP ID
rs10171710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:191255244 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACCCATCCTGTACTGAATCTTTA[A/G]TGTTGAACAAAACAGATGTGGTTTC
Phenotype
MIM: 606537
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1B PubMed Links

Gene Details

Gene
MYO1B
Gene Name
myosin IB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005246572.1 Intron XP_005246629.1
XM_017004156.1 Intron XP_016859645.1
XM_017004157.1 Intron XP_016859646.1
XM_017004158.1 Intron XP_016859647.1

View Full Product Details