Product Details

SNP ID
rs9607567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38690245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGGGGAGAAAGGGAAGTGTGG[C/T]GAGTTCCTCAGGACATAAAGTGCAT
Phenotype
MIM: 615323 MIM: 607046
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
JOSD1 PubMed Links

Gene Details

Gene
JOSD1
Gene Name
Josephin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014876.5 Intron NP_055691.1
XM_005261876.2 Intron XP_005261933.1
XM_005261877.2 Intron XP_005261934.1
XM_005261878.2 Intron XP_005261935.1
XM_005261879.2 Intron XP_005261936.1
Gene
TOMM22
Gene Name
translocase of outer mitochondrial membrane 22
There are no transcripts associated with this gene.

View Full Product Details