Product Details

SNP ID
rs6115611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2700847 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTCAATCAGGAGAAGGAGGCACC[A/G]TCCACACCTGCCTCTGTTGCTCTCG
Phenotype
MIM: 609935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EBF4 PubMed Links

Gene Details

Gene
EBF4
Gene Name
early B-cell factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110514.1 Intron NP_001103984.1
XM_006723600.1 Intron XP_006723663.1
XM_017027983.1 Intron XP_016883472.1
XM_017027984.1 Intron XP_016883473.1
XM_017027985.1 Intron XP_016883474.1

View Full Product Details