Product Details

SNP ID
rs5959856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:81220607 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTTTAGTGTTTTTTTTTCATTT[C/T]AATTTAATTTATTTTTGATCTGCCC
Phenotype
MIM: 300190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3BGRL PubMed Links

Gene Details

Gene
SH3BGRL
Gene Name
SH3 domain binding glutamate rich protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003022.2 Intron NP_003013.1
XM_011531013.1 Intron XP_011529315.1
XM_011531014.1 Intron XP_011529316.1

View Full Product Details