Product Details

SNP ID
rs9803801
Assay Type
Functionally tested
NCBI dbSNP Submissions
45
Location
Chr.1:234634047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGGGTTTGGATGAATCGAATC[C/T]CCTGTTATGCGTGAAATTGTACCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details