Product Details

SNP ID
rs8177080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47625190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCATTCACTCCCATCCCAGTTCT[T/C]TCAATTGCTTCTTCCTCCACTCAGT
Phenotype
MIM: 300383 MIM: 313440
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CFP PubMed Links

Gene Details

Gene
CFP
Gene Name
complement factor properdin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145252.1 Intron NP_001138724.1
NM_002621.2 Intron NP_002612.1
XM_017029575.1 Intron XP_016885064.1
Gene
SYN1
Gene Name
synapsin I
There are no transcripts associated with this gene.

View Full Product Details