Product Details
- SNP ID
-
rs6803590
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:93931664 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCCCTTTCCTTTAGGGCCCAATCG[A/G]ACAGCTACTACTGTTGCTTGCAATA
- Phenotype
-
MIM: 176880
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PROS1
PubMed Links
Gene Details
- Gene
- PROS1
- Gene Name
- protein S (alpha)
View Full Product Details