Product Details
- SNP ID
-
rs10458913
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:44263723 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCATAGCAGGGCCTGGAGGCCGAGC[A/G]TGGCCACACGTCCTCTAGCAAAGCC
- Phenotype
-
MIM: 605420
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ALX4
PubMed Links
Gene Details
- Gene
- ALX4
- Gene Name
- ALX homeobox 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021926.3 |
2471 |
UTR 3 |
|
|
NP_068745.2 |
View Full Product Details