Product Details
- SNP ID
-
rs6043374
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:1629306 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAACCTGGAGAGCGTCTGTGGTTTA[C/T]GGTCAGTCTCAAGGCGATGGATGGG
- Phenotype
-
MIM: 603889
MIM: 605466
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SIRPB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140526422] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SIRPB1
- Gene Name
- signal regulatory protein beta 1
There are no transcripts associated with this gene.
- Gene
- SIRPG
- Gene Name
- signal regulatory protein gamma
- Gene
- SIRPG-AS1
- Gene Name
- SIRPG antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details