Product Details

SNP ID
rs4238109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51096899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAATAATTATGTGCAAGGGTAC[A/G]GGTAGGGAACGAAACAATTTCATCG
Phenotype
MIM: 189889
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFCP2 PubMed Links

Gene Details

Gene
TFCP2
Gene Name
transcription factor CP2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173452.1 Intron NP_001166923.1
NM_001173453.1 Intron NP_001166924.1
NM_005653.4 Intron NP_005644.2

View Full Product Details