Product Details

SNP ID
rs10190853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169816333 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTCGTTACTAGAGAAGTTTCTCC[A/G]AAAGTGTAGAGCACTGTATCATGAA
Phenotype
MIM: 109090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL5 PubMed Links

Gene Details

Gene
METTL5
Gene Name
methyltransferase like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293186.1 Intron NP_001280115.1
NM_001293187.1 Intron NP_001280116.1
NM_014168.3 Intron NP_054887.2
XM_011511023.1 Intron XP_011509325.1
Gene
SSB
Gene Name
Sjogren syndrome antigen B
There are no transcripts associated with this gene.

View Full Product Details