Product Details

SNP ID
rs9463494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49512666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTGTTATAAATGCAGTGAAATTG[C/T]CTTTAAGAGCTGAATTTCTTGAGCA
Phenotype
MIM: 614763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYATL3 PubMed Links

Gene Details

Gene
GLYATL3
Gene Name
glycine-N-acyltransferase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010904.1 Intron NP_001010904.1

View Full Product Details