Product Details

SNP ID
rs6064611
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:38483700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAACAACCCCATGGCAAGTAGA[A/C]TAAGACATGGCAAGTGTGATCAAGA
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
RALGAPB PubMed Links

Gene Details

Gene
RALGAPB
Gene Name
Ral GTPase activating protein non-catalytic beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282917.1 Intron NP_001269846.1
NM_001282918.1 Intron NP_001269847.1
NM_020336.3 Intron NP_065069.1

View Full Product Details