Product Details
- SNP ID
-
rs4602287
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35780106 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGTGCTCTTACATCCCTCTCTCT[C/G]TAGCTAACGTGCTTTTTCCCCTAGA
- Phenotype
-
MIM: 610335
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PHF20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77189140] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PHF20
- Gene Name
- PHD finger protein 20
View Full Product Details