Product Details
- SNP ID
-
rs8178607
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:93935148 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTTGGTCAGTGATCAATGAAGAT[A/G]CTTCATTGAAGGGATTCAAGAGCTC
- Phenotype
-
MIM: 176880
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROS1
PubMed Links
Gene Details
- Gene
- PROS1
- Gene Name
- protein S (alpha)
View Full Product Details