Product Details

SNP ID
rs11902994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99349001 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTGGGCTTAATCAGCTGGAGTTG[G/T]CAGCTGGAGCATCTCTATTTGATCT
Phenotype
MIM: 606086
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF5B PubMed Links

Gene Details

Gene
EIF5B
Gene Name
eukaryotic translation initiation factor 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015904.3 Intron NP_056988.3

View Full Product Details