Product Details
- SNP ID
-
rs12776349
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:48985580 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATGGCCCTGCTGTAGACTCCCGA[A/G]GCATCGGAAGCAGAGTCCGCTGCTG
- Phenotype
-
MIM: 613316
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4294
PubMed Links
Gene Details
- Gene
- MIR4294
- Gene Name
- microRNA 4294
There are no transcripts associated with this gene.
- Gene
- WDFY4
- Gene Name
- WDFY family member 4
There are no transcripts associated with this gene.
View Full Product Details