Product Details
- SNP ID
-
rs13233793
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:148600102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAAAACAGGCTCCAATGGGCACA[A/G]CCTCTTGTCTCTGGGTACCTCTAGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C7orf33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143048176] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C7orf33
- Gene Name
- chromosome 7 open reading frame 33
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145304.3 |
|
Intron |
|
|
NP_660347.1 |
View Full Product Details