Product Details
- SNP ID
-
rs3767209
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:70214530 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- GGTCACAGATAGATTTTTGCCTATA[A/G]TTTTAGGGTGTTCACAGACCCTGCT
- Phenotype
-
MIM: 602010
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143424791] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC40
- Gene Name
- leucine rich repeat containing 40
There are no transcripts associated with this gene.
- Gene
- SRSF11
- Gene Name
- serine and arginine rich splicing factor 11
View Full Product Details