Product Details

SNP ID
rs13395505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190973788 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCAACAACAATCAGGAAAGTGTA[A/G]GAAGGTACAATCTCTAATGATTGTA
Phenotype
MIM: 138280 MIM: 600555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLS PubMed Links

Gene Details

Gene
GLS
Gene Name
glutaminase
There are no transcripts associated with this gene.

Gene
STAT1
Gene Name
signal transducer and activator of transcription 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007315.3 Intron NP_009330.1
NM_139266.2 Intron NP_644671.1
XM_006712718.1 Intron XP_006712781.1
XM_017004783.1 Intron XP_016860272.1

View Full Product Details