Product Details

SNP ID
rs10804578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126577573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGATAAAGGGGAAGGGAAAGGG[C/T]TGCGGGCAGAGAAAGTGCAGAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNRD3NB PubMed Links

Gene Details

Gene
TXNRD3NB
Gene Name
thioredoxin reductase 3 neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039783.1 Intron NP_001034872.1

View Full Product Details