Product Details

SNP ID
rs12076179
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:41508813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCACACAGCCTGGCTGAGGACTG[A/G]GGATTAGCCAGAGAGGTCCCTGCTC
Phenotype
MIM: 606649
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIVEP3 PubMed Links

Gene Details

Gene
HIVEP3
Gene Name
human immunodeficiency virus type I enhancer binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127714.2 9871 UTR 3 NP_001121186.1
NM_024503.4 9871 UTR 3 NP_078779.2
XM_011541884.2 9871 UTR 3 XP_011540186.1
XM_011541885.2 9871 Intron XP_011540187.1
XM_011541886.2 9871 Intron XP_011540188.1
XM_011541887.2 9871 Intron XP_011540189.1
XM_017001992.1 9871 UTR 3 XP_016857481.1
XM_017001993.1 9871 UTR 3 XP_016857482.1
XM_017001994.1 9871 UTR 3 XP_016857483.1

View Full Product Details