Product Details

SNP ID
rs12143919
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:219182046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGTTTATGCCTATGTGAGGTTT[A/T]AAAAAGTGTATTTTGTTGTGGTAAA
Phenotype
MIM: 616548
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LYPLAL1 PubMed Links

Gene Details

Gene
LYPLAL1
Gene Name
lysophospholipase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300769.1 Intron NP_001287698.1
NM_001300770.1 Intron NP_001287699.1
NM_001300771.1 Intron NP_001287700.1
NM_001300772.1 Intron NP_001287701.1
NM_138794.4 Intron NP_620149.2
XM_006711162.3 Intron XP_006711225.1
XM_011509182.2 Intron XP_011507484.1
XM_011509183.2 Intron XP_011507485.1
XM_017000271.1 Intron XP_016855760.1
XM_017000272.1 Intron XP_016855761.1
Gene
LYPLAL1-AS1
Gene Name
LYPLAL1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details